EMAIL THIS PAGE TO A FRIEND
417610 Aldrich

N-Propylphthalimide

98%

  • CAS Number 5323-50-2

  • Empirical Formula (Hill Notation) C11H11NO2

  • Molecular Weight 189.21

  •  MDL number MFCD00014584

Purchase

Properties

Related Categories Amines, Building Blocks, C11, C11 to C12, Carbonyl Compounds,
InChI Key   RLARUBPTQYKZKA-UHFFFAOYSA-N
assay   98%

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?