EMAIL THIS PAGE TO A FRIEND
41763 Aldrich

3,9-Dimethylxanthine

≥99.0%

Synonym: 2,6-Dihydroxy-3,9-dimethylpurine

  • CAS Number 15837-08-8

  • Empirical Formula (Hill Notation) C7H8N4O2

  • Molecular Weight 180.16

  •  Beilstein Registry Number 3679243

  •  EC Number 239-942-4

  •  MDL number MFCD00042777

  •  PubChem Substance ID 57650088

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Heterocyclic Building Blocks, Purines
InChI Key   HEOWZFHIJSYJIC-UHFFFAOYSA-N
assay   ≥99.0%
mp   ≥300 °C
Gene Information   rat ... Adora1(29290), Adora2a(25369)

Description

Packaging

250 mg in glass bottle

Bottomless glass bottle. Contents are inside inserted fused cone.

Application

3,9-Dimethylxanthine has been used to study the molecular mechanism of in situ activation of ryanodine receptors (RY) in pancreatic β cells from ob/ob mice.

General description

3,9-Dimethylxanthine is a xanthine derivative. It has been reported to be an activator of ryanodine receptor (RY). Mechanism of its electrochemical oxidation has been investigated in the pH range 3.0-10.8 at stationary pyrolytic graphite electrode. It has been synthesized by reacting 1-methyl-5-(methylamino)-imidazole-4-carboxamide with EtOCOCl in acetate buffer (pH5), followed by the reaction with aqueous NaOH.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?