• USA Home
  • 417998 - Poly(hexamethylene diisocyanate)

EMAIL THIS PAGE TO A FRIEND
417998 Aldrich

Poly(hexamethylene diisocyanate)

viscosity 5,000-10,000 cP (25 °C)(lit.)

Purchase

Properties

Related Categories Engineering Polymers, Materials Science, Polymer Science, Polymers, Polyurethanes and Urethane Precursors More...
refractive index   n20/D 1.505
viscosity   5,000-10,000 cP(25 °C)(lit.)
density   1.14 g/mL at 25 °C

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
WGK Germany 
3
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

comparable product

Product #

Description

Add to Cart

418005 Poly(hexamethylene diisocyanate), viscosity 1,300-2,200 cP (25 °C)(lit.)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?