• USA Home
  • 41844 - (±)-3-(Boc-amino)-4-(4-biphenylyl)butyric acid

EMAIL THIS PAGE TO A FRIEND
41844 Aldrich

(±)-3-(Boc-amino)-4-(4-biphenylyl)butyric acid

≥98.0% (HPLC)

Synonym: Boc-4′-phenyl-DL-β-homophenylalanine

Purchase

Properties

Related Categories Beta-Amino Acids, Chemical Biology, Chemical Synthesis, Peptide Chemistry, Unnatural Amino Acids & Derivatives More...
InChI Key   SHRZMJHRBQEGBC-UHFFFAOYSA-N
assay   ≥98.0% (HPLC)
storage temp.   2-8°C

Description

Packaging

500 mg in poly tube

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Articles

β-Amino Acids and Homologs

Although they are less abundant than their α-analogues, β-amino acids occur in nature both in free form and bound to peptides. Oligomers composed exclusively of β-amino acids (so-called β-peptides) m...
Matthias Junkers
Aldrich ChemFiles 2008, 8.7, 11.
Keywords: Chemfiles, Degradations, Metabolism, Peptidomimetics

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?