• USA Home
  • 418757 - Pentaerythritol propoxylate (17/8 PO/OH)

EMAIL THIS PAGE TO A FRIEND
418757 Aldrich

Pentaerythritol propoxylate (17/8 PO/OH)

average Mn ~629

Purchase

Properties

Related Categories Crosslinkers, Crosslinking Agents, Materials Science, Polymer Science, Reagents for Polymerization More...
mol wt   average Mn ~629
refractive index   n20/D 1.46
bp   >300 °C(lit.)
density   1.05 g/mL at 25 °C(lit.)

Description

Packaging

500 mL in poly bottle

Price and Availability

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
1
Flash Point(F) 
377.6 °F
Flash Point(C) 
192 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?