• USA Home
  • 418838 - tert-Amyl-tert-butyl nitroxide

EMAIL THIS PAGE TO A FRIEND
418838 Aldrich

tert-Amyl-tert-butyl nitroxide

95%

Purchase

Properties

Related Categories Chemical Synthesis, Other Synthetic Reagents for Radical Chemistry, Radical Chemistry, Synthetic Reagents
InChI Key   RRUMSABVDBCPNH-UHFFFAOYSA-N
assay   95%
refractive index   n20/D 1.441(lit.)
bp   55-56 °C/5 mmHg(lit.)
density   0.86 g/mL at 25 °C(lit.)
storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 1993C 3 / PGIII
WGK Germany 
3
Flash Point(F) 
134.6 °F
Flash Point(C) 
57 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?