• USA Home
  • 418986 - Aluminum 2,9,16,23-tetraphenoxy-29H,31H-phthalocyanine hydroxide

EMAIL THIS PAGE TO A FRIEND
418986 Aldrich

Aluminum 2,9,16,23-tetraphenoxy-29H,31H-phthalocyanine hydroxide

Dye content 90 %

Purchase

Properties

Related Categories Infrared (IR) Dyes, Materials Science, Organic and Printed Electronics, Photonic and Optical Materials, Phthalocyanine and Porphyrin Dyes,
InChI Key   DZXAWAFIWWBJOI-UHFFFAOYSA-M
composition   Dye content, 90%
absorption   λmax 681 nm
  λmax 686 nm (secondary)

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?