• USA Home
  • 420115 - Tetraethylammonium trifluoroacetate

EMAIL THIS PAGE TO A FRIEND
420115 Aldrich

Tetraethylammonium trifluoroacetate

98%

Synonym: Trifluoroacetic acid tetraethylammonium salt

  • CAS Number 30093-29-9

  • Linear Formula (CH3CH2)4N(CF3COO)

  • Molecular Weight 243.27

  •  Beilstein Registry Number 6254213

  •  MDL number MFCD00192470

  •  PubChem Substance ID 24866312

  • Popular Documents:  FTIR (PDF)  

Purchase

Properties

Related Categories Ammonium Salts, Chemical Synthesis, Phase Transfer Catalysts, Synthetic Reagents
InChI Key   SBOOKGHQWGEWCB-UHFFFAOYSA-M
assay   98%
mp   74-76 °C(lit.)

Description

Packaging

1 g in glass bottle

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3263 8 / PGII
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?