• USA Home
  • 420379 - 1,1,3,3,3-Pentakis(dimethylamino)-1λ5,3λ5-diphosphazene 1-oxide

EMAIL THIS PAGE TO A FRIEND
420379 Aldrich

1,1,3,3,3-Pentakis(dimethylamino)-5,3λ5-diphosphazene 1-oxide

98%

Synonym: PDPO

  • CAS Number 91241-12-2

  • Linear Formula [(CH3)2N]3P=NP(O)[N(CH3)2]2

  • Molecular Weight 312.33

  •  Beilstein Registry Number 1886624

  •  MDL number MFCD00192491

  •  PubChem Substance ID 24866333

Purchase

Properties

Related Categories Acids & Bases, Bases, Chemical Synthesis, Phosphazene Bases, Synthetic Reagents More...
InChI Key   RVZIHXHEDQCEED-UHFFFAOYSA-N
assay   98%
refractive index   n20/D 1.495(lit.)
bp   348 °C(lit.)
density   1.07 g/mL at 25 °C(lit.)

Description

Packaging

1 g in glass bottle

Bottomless glass bottle. Contents are inside inserted fused cone.

Application

Phosphazene Bases: a Family of Extremely Strong, Non-Ionic, Non-Charged Nitrogen–Bases

Price and Availability


Cross-Coupling Guide
Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3267 8 / PGII
WGK Germany 
3
Flash Point(F) 
210.2 °F
Flash Point(C) 
99 °C
Protocols & Articles

Related Content

Molarity Calculator and Normality Calculator for Acids and Bases

The molarity calculator tool provides lab-ready directions describing how to prepare an acid or base solution of specified Molarity (M) or Normality (N) from a concentrated acid or base solution.  To...

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?