• USA Home
  • 420506 - Dicyanocobyrinic acid heptamethyl ester

EMAIL THIS PAGE TO A FRIEND
420506 Aldrich

Dicyanocobyrinic acid heptamethyl ester

97%

Synonym: Heptamethyl dicyano cobyrinate

  • CAS Number 36522-80-2

  • Empirical Formula (Hill Notation) C54H73CoN6O14

  • Molecular Weight 1089.12

  •  Beilstein Registry Number 1207601

  •  MDL number MFCD00192013

Purchase

Properties

InChI Key   CPBDBBXBECBXRW-NTQATXMTSA-N
assay   97%
mp   124-125 °C (dec.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?