• USA Home
  • 421111 - VOLAN® L bonding agent

EMAIL THIS PAGE TO A FRIEND
421111 Aldrich

VOLAN® L bonding agent

Purchase

Properties

refractive index   n20/D 1.402(lit.)
bp   77 °C(lit.)
density   0.95 g/mL at 25 °C(lit.)

Description

Legal Information

DuPont Product

VOLAN is a registered trademark of Zaclon LLC

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 1993BE 3 / PGII
WGK Germany 
1
Flash Point(F) 
1.4 °F
Flash Point(C) 
-17 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?