• USA Home
  • 421294 - MERPOL® DA surfactant

EMAIL THIS PAGE TO A FRIEND
421294 Aldrich

MERPOL® DA surfactant

60 wt. % in water: isobutanol (ca. 50:50)

Purchase

Properties

Related Categories Materials Science, Micro/NanoElectronics, Nonionic, Nonionic Surfactants, Polymer Additives,
concentration   60 wt. % in water: isobutanol (ca. 50:50)
refractive index   n20/D 1.429(lit.)
bp   100 °C(lit.)
density   1.03 g/mL at 25 °C(lit.)

Description

Packaging

250 mL in glass bottle

Legal Information

MERPOL is a registered trademark of Stepan Co.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN1212 - class 3 - PG 3 - Isobutanol
WGK Germany 
3
Flash Point(F) 
114.8 °F
Flash Point(C) 
46 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?