• USA Home
  • 421332 - MERPOL® OJ surfactant

EMAIL THIS PAGE TO A FRIEND
421332 Aldrich

MERPOL® OJ surfactant

Purchase

Properties

Related Categories Materials Science, Micro/NanoElectronics, Nonionic, Nonionic Surfactants, Polymer Additives,
refractive index   n20/D 1.469(lit.)
solubility   water and organic: soluble
density   1 g/mL at 25 °C(lit.)
HLB   12

Description

Application

Nonionic, alcohol ethoxylate. Wetting, dispersing, and emulsifying agent.

Packaging

1 L in poly bottle

Legal Information

MERPOL is a registered trademark of Stepan Co.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
2
Flash Point(F) 
302 °F
Flash Point(C) 
150 °C

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?