• USA Home
  • 421421 - Zonyl® FSN-100 fluorosurfactant

EMAIL THIS PAGE TO A FRIEND
421421 Aldrich

Zonyl® FSN-100 fluorosurfactant

Synonym: Zonyl® fluorosurfactant

  • CAS Number 65545-80-4

  • Linear Formula (C2H4O)x(CF2)yC2H5FO

Purchase

Description

Legal Information

Zonyl is a registered trademark of E. I. du Pont de Nemours and Company

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?