EMAIL THIS PAGE TO A FRIEND
421456 Aldrich

Zonyl® FSO-100

Synonym: Zonyl® fluorosurfactant

  • CAS Number 65545-80-4

  • Linear Formula (C2H4O)x(CF2)yC2H5FO

Purchase

Properties

Related Categories Materials Science, Micro/NanoElectronics, Nonionic, Nonionic Surfactants, Polymer Additives,
refractive index   n20/D 1.3950
bp   200 °C
density   1.36 g/mL at 25 °C

Description

Legal Information

Zonyl is a registered trademark of E. I. du Pont de Nemours and Company

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?