• USA Home
  • 422290 - α-Oxo-N-[(R)-1-phenylethyl]phenylacetamide

EMAIL THIS PAGE TO A FRIEND
422290 Aldrich

α-Oxo-N-[(R)-1-phenylethyl]phenylacetamide

98%

Purchase

Properties

Related Categories Amides, Asymmetric Synthesis, Chemical Synthesis, Chiral Building Blocks, Organic Building Blocks More...
InChI Key   KCDFERKGOUXJDD-GFCCVEGCSA-N
assay   98%
optical activity   [α]20/D +105°, c = 2 in ethanol
mp   110-112 °C(lit.)

Description

Packaging

1 g in glass bottle

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?