• USA Home
  • 42544 - Dimethyl (4S,5S)-1,3,2-dioxathiolane-4,5-dicarboxylate 2,2-dioxide

EMAIL THIS PAGE TO A FRIEND
42544 Aldrich

Dimethyl (4S,5S)-1,3,2-dioxathiolane-4,5-dicarboxylate 2,2-dioxide

Synonym: (+)-Dimethyl 2,3-O-sulfonyl-D-tartrate, Dimethyl D-tartrate cycl-sulfate

  • CAS Number 127854-46-0

  • Empirical Formula (Hill Notation) C6H8O8S

  • Molecular Weight 240.19

  •  Beilstein Registry Number 6219088

  •  MDL number MFCD00077753

  •  PubChem Substance ID 24866634

Purchase

Properties

Related Categories Asymmetric Synthesis, Chemical Synthesis, Chiral Building Blocks, Esters, Organic Building Blocks More...
InChI Key   YZPCWPMIVKWDOZ-IMJSIDKUSA-N
optical activity   [α]20/D +73±2°, c = 2% in chloroform
mp   68-71 °C
storage temp.   2-8°C

Description

Packaging

1 g in glass bottle

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?