• USA Home
  • 425605 - 4,6-Dihydroxy-2,5-diphenylpyrimidine

EMAIL THIS PAGE TO A FRIEND
425605 Aldrich

4,6-Dihydroxy-2,5-diphenylpyrimidine

97%

Synonym: 2,5-Diphenyl-4,6-pyrimidinediol

  • CAS Number 29133-86-6

  • Empirical Formula (Hill Notation) C16H12N2O2

  • Molecular Weight 264.28

  •  MDL number MFCD00216578

  •  PubChem Substance ID 24866648

Purchase

Properties

Related Categories Building Blocks, C12 to C30+, Chemical Synthesis, Heterocyclic Building Blocks, Pyrimidines More...
InChI Key   XYXQJMOZTJTMBV-UHFFFAOYSA-N
assay   97%
mp   >300 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?