• USA Home
  • 426652 - Poly(ethylene-co-methacrylic acid)

EMAIL THIS PAGE TO A FRIEND
426652 Aldrich

Poly(ethylene-co-methacrylic acid)

pellets, melt index 35 g/10 min (190°C/2.16kg)

  • CAS Number 25053-53-6

  • Linear Formula (CH2CH2)x[CH2C(CH3)(CO2H)]y

Purchase

Properties

form   pellets
melt index   35 g/10 min (190°C/2.16kg)
tensile impact   723 kJ/m2± (23°C)(ASTM D 1822S)
mp   76 °C(lit.)
transition temp   softening point 75 °C (Vicat, ASTM D 1525, rate B)

Description

Physical form

Branched, random co-polymer.

Application

Molded or extruded parts requiring flexibility, tensile strength and low temperature impact and adhesion.

Features and Benefits

Tough, light-weight, easily processed, inherent flexibility without need for plasticizers. Excellent adhesion properties.

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?