• USA Home
  • 426687 - Poly(ethylene-co-methacrylic acid) zinc salt

EMAIL THIS PAGE TO A FRIEND
426687 Aldrich

Poly(ethylene-co-methacrylic acid) zinc salt

melt index 1.1 g/10 min (190°C/2.16kg)

  • Linear Formula (-CH2CH2-)x[-CH2C(CH3)(CO2R)-]y, R = H or Zn/2

  •  MDL number MFCD00212544

  •  PubChem Substance ID 24866718

Purchase

Properties

Related Categories Ethylene, Hydrophobic Polymers, Materials Science, Olefins, Polymer Science,
melt index   1.1 g/10 min (190°C/2.16kg)
composition   methacrylic acid, 10 wt. %
tensile impact   1190 kJ/m2± (23°C)(ASTM D 1822S)
mp   82 °C
transition temp   softening point 74 °C (Vicat, ASTM D 1525-70)
density   0.95 g/mL at 25 °C

Description

Application

Golf ball covers, auto rub strips, athletic shoe soles, ski boots and cosmetic packaging.

Features and Benefits

Excellent impact strength and optical properties. Foamable, abrasion and solvent resistant and exhibits adhesion to various materials.

Price and Availability


Webinar: Recent Developments in RAFT Agents
Safety & Documentation

Safety Information

WGK Germany 
-

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?