• USA Home
  • 426989 - Poly(styrene-co-divinylbenzene)

EMAIL THIS PAGE TO A FRIEND
426989 Aldrich

Poly(styrene-co-divinylbenzene)

mean pore size 46 Å, 300-800 μm particle size

Synonym: Polystyrene crosslinked with divinylbenzene

Purchase

Properties

Related Categories Adsorbent Media (Polymeric), Amberlite, Analytical/Chromatography, Anion, LPLC Separation Media,
impurities   <5% water
matrix   Poly(styrene-co-divinylbenzene)
particle size   300-800 μm
pore size   46 Å mean pore size
surface area   1,100 m2/g

Description

Application

Adsorbent to concentrate organics from water.

Physical form

Macroporous polymeric adsorbent spheres,

Features and Benefits

Highly cross-linked polymer bead. Very versatile adsorbent, may have low capacity for smaller organic molecules.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles

Articles

Selection Guide for Polymeric Resins in Chromatography & Purification

Sigma-Aldrich carries a vast array of resins, from leading manufacturers; such as, Dow Chemical, Rohm & Haas, Mitsubishi, and Lanxess. Our product line includes adsorbent, ion-exchange (anion / catio...
Keywords: Adsorption, Antibiotics, Detergents, Fermentation, Filtration, Hormones, Ion Exchange, Pharmaceutical, Purification, Separation

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?