EMAIL THIS PAGE TO A FRIEND
428000 Aldrich

Polyethylene

pellets

Purchase

Properties

form   pellets
melt index   40 g/10 min (190°C/2.16kg)
hardness   68 (Shore D, ASTM D 2240)
transition temp   softening point 122 °C (Vicat, ASTM D 1525)
density    at 25 °C (high density)

Description

Application

Injection molded overcaps, thin-walled containers, toys and tubs.

Features and Benefits

Excellent flow and warp resistance.

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?