EMAIL THIS PAGE TO A FRIEND
428019 Aldrich

Polyethylene

High density, melt index 42 g/10 min (190°C/2.16kg)

Purchase

Properties

Related Categories Ethylene, Hydrophobic Polymers, Materials Science, Olefins, Polymer Science,
form   pellets
melt index   42 g/10 min (190°C/2.16kg)
impact strength   29.9 J/m (Izod, ASTM D 256, −50 °C)
mp   121 °C
transition temp   Tm (DSC) 121 °C (onset)
density   0.947 g/mL at 25 °C

Description

Application

Injection molded, thin-walled containers and housewares.

Features and Benefits

High stiffness and impact and warp resistance.

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?