• USA Home
  • 428353 - Poly(ethylene-co-carbon monoxide)

EMAIL THIS PAGE TO A FRIEND
428353 Aldrich

Poly(ethylene-co-carbon monoxide)

average Mn ~3,000

Purchase

Properties

InChI Key   INVGSXKPOIHXPB-UHFFFAOYSA-N
mol wt   average Mn ~3,000
contains   polar carbonyl group as adhesion promoter
hardness   7 dmm (Penetrometer, ASTM D 5)
viscosity   230 cP(140 °C, Brookfield)(lit.)
transition temp   Tg ~−40 °C
  Tm 85-90 °C

Description

Application

Additive to formulated adhesives.

Features and Benefits

Improves adhesion, flexibility, elevated service temperature and set temperature and gives lower viscosity to adhesives.

Physical form

Synthetic ketone wax.

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?