• USA Home
  • 429511 - 1-(3-Chlorophenyl)-3-methyl-2-pyrazolin-5-one

EMAIL THIS PAGE TO A FRIEND
429511 Aldrich

1-(3-Chlorophenyl)-3-methyl-2-pyrazolin-5-one

97%

Purchase

Properties

Related Categories Building Blocks, C10, C10-C30+, Chemical Synthesis, Halogenated Heterocycles,
InChI Key   RIOMUJXIGYZENC-UHFFFAOYSA-N
assay   97%
mp   128-131 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
2

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?