• USA Home
  • 430277 - Glycidyl 4-nonylphenyl ether

EMAIL THIS PAGE TO A FRIEND
430277 Aldrich

Glycidyl 4-nonylphenyl ether

mol wt 300-325 g/epoxide by perchloric acid method, technical grade

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Epoxides, Organic Building Blocks, Oxygen Compounds More...
grade   technical grade
InChI Key   AVKQYWUBGXNBCW-UHFFFAOYSA-N
mol wt   mol wt 300-325 g/epoxide by perchloric acid method
impurities   ≤10 ppm free epichlorohydrin
refractive index   n20/D 1.509(lit.)
viscosity   90-130 cSt(25 °C, Ubbelohde)(lit.)
bp   312 °C(lit.)
density   0.978 g/mL at 25 °C(lit.)

Description

Packaging

100 mL in glass bottle

Application

Glycidyl 4-nonylphenyl ether may be employed for the preparation of patterned template for the surface patterning of Escherichia coli (a gram negative bacterium) and Bacillus subtilis (a gram positive bacterium).

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?