• USA Home
  • 432806 - 4-Methoxy-3-biphenylamine hydrochloride

EMAIL THIS PAGE TO A FRIEND
432806 Aldrich

4-Methoxy-3-biphenylamine hydrochloride

99%

Purchase

Properties

Related Categories Amine Salts, Building Blocks, Chemical Synthesis, Nitrogen Compounds, Organic Building Blocks More...
InChI Key   QLWYZBIVFJWGMG-UHFFFAOYSA-N
assay   99%
mp   227-230 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?