EMAIL THIS PAGE TO A FRIEND
432814 Aldrich

Acetonitrile-d3

99.8 atom % D

Synonym: Methyl-d3 cyanide, Trideuteroacetonitrile

Purchase

Properties

InChI Key   WEVYAHXRMPXWCK-FIBGUPNXSA-N
isotopic purity   99.8 atom % D
refractive index   n20/D 1.341(lit.)
bp   80.7 °C(lit.)
density   0.844 g/mL at 25 °C(lit.)
mass shift   M+3

Description

Packaging

10mL unit packaged in ampules, 50mL unit packaged in screw-cap bottle

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 1648 3 / PGII
WGK Germany 
2
Flash Point(F) 
42.8 °F
Flash Point(C) 
6 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?