• USA Home
  • 433721 - (1R)-trans-N,N′-1,2-Cyclohexanediylbis(1,1,1-trifluoromethanesulfonamide)

EMAIL THIS PAGE TO A FRIEND
433721 Aldrich

(1R)-trans-N,N′-1,2-Cyclohexanediylbis(1,1,1-trifluoromethanesulfonamide)

97%

Purchase

Properties

Related Categories Asymmetric Synthesis, Chemical Synthesis, Chiral Building Blocks, Organic Building Blocks, Sulfonamides/Sulfinamides More...
InChI Key   GKSGSDYYIYURPD-PHDIDXHHSA-N
assay   97%
optical activity   [α]20/D −6.1°, c = 5 in ethanol
mp   185-187 °C(lit.)

Description

Packaging

1 g in glass bottle

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?