Synonym: Hexadeuterobenzene
CAS Number 1076-43-3
Empirical Formula (Hill Notation) C6D6
Molecular Weight 84.15
Beilstein Registry Number 1905426
EC Number 214-061-8
MDL number MFCD00003010
PubChem Substance ID 24867198
Popular Documents: Specification Sheet (PDF) | FTNMR (PDF)
| Related Categories | Aldrich 100% NMR Solvents for High Resolution, Alphabetical Listings, B, Benzene-d6, High Resolution NMR, |
| isotopic purity | 99.96 atom % D |
| packaging | pk of 10 × 1 mL |
| refractive index |
n |
| bp | 79.1 °C/760 mmHg(lit.) |
| mp | 6.8 °C(lit.) |
| density | 0.950 g/mL at 25 °C(lit.) |
| mass shift | M+6 |
1 pkg in ampule
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?