• USA Home
  • 436445 - 8-Hydroxyquinoline copper(II) salt

EMAIL THIS PAGE TO A FRIEND
436445 Aldrich

8-Hydroxyquinoline copper(II) salt

98%

Purchase

Properties

InChI Key   YXLXNENXOJSQEI-UHFFFAOYSA-L
assay   98%
mp   240 °C (dec.)(lit.)

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?