• USA Home
  • 43653 - 2,3-Di(4-pyridyl)-2,3-butanediol

EMAIL THIS PAGE TO A FRIEND
43653 Aldrich

2,3-Di(4-pyridyl)-2,3-butanediol

mixture of racemic and meso forms, ≥98.0% (CHN)

  • CAS Number 69267-29-4

  • Empirical Formula (Hill Notation) C14H16N2O2

  • Molecular Weight 244.29

  •  Beilstein Registry Number 201002

  •  MDL number MFCD00043009

  •  PubChem Substance ID 57650149

Purchase

Properties

Related Categories Building Blocks, C13 to C15, Chemical Synthesis, Heterocyclic Building Blocks, Pyridines More...
InChI Key   GBOSNRFNGIFXLO-UHFFFAOYSA-N
assay   ≥98.0% (CHN)
mp   220-222 °C
solubility   5 M HCl: soluble0.1 g/10 mL, clear, colorless
storage temp.   2-8°C

Description

Packaging

5 g in glass bottle

Application

2,3-Di(4-pyridyl)-2,3-butanediol (dpb) may be employed for the preparation of mercury(II) coordination polymers.

General description

2,3-Di(4-pyridyl)-2,3-butanediol (dpb) participates as a ligand in the preparation of IB (group 11) metal thiocyanate based coordination polymers. Post synthetic modification of Zn-based 1,2,4,5-tetrakis(4′-carboxyphenyl) benzene, 2,3-di(4-pyridyl)-2,3-butanediol pillared metal organic framework (MOF) with lithium or magnesium ions has been reported to increase the affinity and uptake of hydrogen.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Articles

Selected Applications of Metal-Organic Frameworks in Sustainable Energy Technologies

Although the field of high surface area materials is considered very mature, the need for improved materials for alternative energy applications remains pressing. Currently, zeolites, metal oxides, a...
Stephen R. Caskey, Adam J. Matzger
Material Matters 2009, 4.4, 111.
Keywords: Adsorption, Alternative energy, Applications, Capabilities, Combustion, Diffraction, Environmental, Filtration, Infrared spectroscopy, Ligands, Metal organic frameworks, Purification, Scintillation, Separation, Solvents, Sonication, Spectra, Tools, Type, X-Ray diffraction

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?