• USA Home
  • 436739 - Amberjet® 1200 hydrogen form

EMAIL THIS PAGE TO A FRIEND
436739 Aldrich

Amberjet® 1200 hydrogen form

hydrogen form

Purchase

Properties

Related Categories Amberlite, Amberlite Strong Cation Exchangers, Analytical/Chromatography, Cation, Cation Exchange Media,
moisture   49-55%
impurities   50%
matrix   styrene-divinylbenzene (gel)
particle size   550-650 mesh
capacity   1.8 meq/mL by wetted bed volume

Description

Features and Benefits

Strongly acidic cation exchanger with sulfonic acid functionality, hydrogen form useful in water treatment.

Legal Information

Amberjet is a registered trademark of The Dow Chemical Company or an affiliated company of Dow

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles

Articles

Selection Guide for Polymeric Resins in Chromatography & Purification

Sigma-Aldrich carries a vast array of resins, from leading manufacturers; such as, Dow Chemical, Rohm & Haas, Mitsubishi, and Lanxess. Our product line includes adsorbent, ion-exchange (anion / catio...
Keywords: Adsorption, Antibiotics, Detergents, Fermentation, Filtration, Hormones, Ion Exchange, Pharmaceutical, Purification, Separation

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?