EMAIL THIS PAGE TO A FRIEND
437735 Aldrich

1,4-Dioxane-d8

99 atom % D, contains 0.03 % (v/v) TMS

Synonym: p-Dioxane-d8, Octadeuterodioxane

Purchase

Properties

Related Categories Alphabetical Listings, D, Stable Isotopes More...
InChI Key   RYHBNJHYFVUHQT-SVYQBANQSA-N
isotopic purity   99 atom % D
assay   99% (CP)
contains   0.03 % (v/v) TMS
refractive index   n20/D 1.4198(lit.)
bp   99 °C(lit.)
density   1.129 g/mL at 25 °C(lit.)
mass shift   M+8

Price and Availability


Stable Isotope
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Supplemental Hazard Statements 
May form explosive peroxides., Repeated exposure may cause skin dryness or cracking.
RIDADR 
UN1165 - class 3 - PG 2 - Dioxane
WGK Germany 
3
Flash Point(F) 
41 °F
Flash Point(C) 
5 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?