• USA Home
  • 438847 - 1-(Phenylsulfonyl)-2-pyrrolecarboxaldehyde

EMAIL THIS PAGE TO A FRIEND
438847 Aldrich

1-(Phenylsulfonyl)-2-pyrrolecarboxaldehyde

97%

Purchase

Properties

Related Categories Aldehydes, Building Blocks, C10-C12, Carbonyl Compounds, Chemical Synthesis,
InChI Key   XZMIDORMXBMWTQ-UHFFFAOYSA-N
assay   97%
mp   81-82 °C(lit.)

Description

Packaging

1, 5 g in glass bottle

General description

1-(Phenylsulfonyl)-2-pyrrolecarboxaldehyde is a heterocyclic building block.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?