• USA Home
  • 438855 - 1-(Phenylsulfonyl)-2-pyrrolecarbonitrile

EMAIL THIS PAGE TO A FRIEND
438855 Aldrich

1-(Phenylsulfonyl)-2-pyrrolecarbonitrile

98%

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Heterocyclic Building Blocks, Pyrroles
InChI Key   XXJCLUNFWADISV-UHFFFAOYSA-N
assay   98%
mp   93-96 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?