Synonym: 1-Naphthylacetic anhydride
CAS Number 5415-58-7
Linear Formula (C10H7CH2CO)2O
Molecular Weight 354.40
Beilstein Registry Number 2168120
MDL number MFCD00210070
PubChem Substance ID 24867484
| Related Categories | Analytical Reagents, Analytical/Chromatography, Biochemicals and Reagents, Building Blocks, Carbohydrate Labels, |
| assay | 96% |
| mp | 116-119 °C(lit.) |
| fluorescence | λex 216 nm; λem 360 nm (polyamin adduct) |
| storage temp. | 2-8°C |
1 g in glass bottle
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?