• USA Home
  • 438979 - (R)-(−)-γ-Trityloxymethyl-γ-butyrolactone

EMAIL THIS PAGE TO A FRIEND
438979 Aldrich

(R)-(−)-γ-Trityloxymethyl-γ-butyrolactone

98%

Synonym: (R)-(−)-Dihydro-5-trityloxymethyl-2(3H)-furanone, 5-O-Trityl-2,3-dideoxy-L-glyceropentono-1,4-lactone

  • CAS Number 78158-90-4

  • Empirical Formula (Hill Notation) C24H22O3

  • Molecular Weight 358.43

  •  Beilstein Registry Number 4911276

  •  MDL number MFCD00043001

  •  PubChem Substance ID 24867486

Purchase

Properties

Related Categories Asymmetric Synthesis, Chemical Synthesis, Chiral Building Blocks, Lactones, Organic Building Blocks More...
InChI Key   OZWWLABCDGFABG-JOCHJYFZSA-N
assay   98%
optical activity   [α]20/D −26°, c = 1 in methylene chloride
mp   150-152 °C(lit.)

Description

Packaging

1 g in glass bottle

Application

This compound was used in the synthesis of target pentonolactones for interaction studies of γ-lactones with the phorbol ester receptor of protein kinase

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?