• USA Home
  • 438995 - Microtorch®, Model MT-11 with fuel cell

EMAIL THIS PAGE TO A FRIEND
438995 Aldrich

Microtorch®, Model MT-11 with fuel cell

pkg of 1 ea

Synonym: mini torches, torch lighters, torch micro

Purchase

Properties

Related Categories Burners, Heating, Labware More...
packaging   pkg of 1 ea

Description

General description

Fuel cell comes empty, and does not contain butane.

Legal Information

Ultratorch is a registered trademark of Master Appliance Corp.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

replacement part

Product #

Description

Add to Cart

439002 Micro Ultratorch® fuel cell, pkg of 2 ea

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?