• USA Home
  • 439002 - Micro Ultratorch® fuel cell

EMAIL THIS PAGE TO A FRIEND
439002 Aldrich

Micro Ultratorch® fuel cell

pkg of 2 ea

Synonym: mini torches, torch lighters, torch micro

Purchase

Properties

Related Categories Burners, Heating, Labware More...
packaging   pkg of 2 ea

Description

General description

Fuel cell comes empty, and does not contain butane.

Legal Information

Ultratorch is a registered trademark of Master Appliance Corp.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?