Synonym: (Methyl sulfoxide)-d6, DMSO-d6, Hexadeuterodimethyl sulfoxide
CAS Number 2206-27-1
Linear Formula (CD3)2SO
Molecular Weight 84.17
Beilstein Registry Number 1237248
EC Number 218-617-0
MDL number MFCD00002090
PubChem Substance ID 24867320
Popular Documents: Specification Sheet (PDF) | FTNMR (PDF)
| Related Categories | Aldrich High Purity NMR Solvents for Routine NMR, Alphabetical Listings, D, Dimethyl sulfoxide-d6, High Throughput NMR, |
| vapor pressure | 0.42 mmHg ( 20 °C) |
| isotopic purity | 99.9 atom % D |
| autoignition temp. | 573 °F |
| contains | 0.1 % (v/v) TMS |
| expl. lim. | 42 % |
| refractive index |
n |
| bp | 189 °C(lit.) |
10 g in ampule
25, 50 g in glass bottle
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?