EMAIL THIS PAGE TO A FRIEND
441333 Aldrich

Chloroform-d

99.8 atom % D

Synonym: Deuterochloroform

Purchase

Properties

Related Categories Chloroform, Chloroform-d, NMR Solvents, Solvent by Type, Solvent by Application,
InChI Key   HEDRZPFGACZZDS-MICDWDOJSA-N
isotopic purity   99.8 atom % D
packaging   pk of 10 × 0.75 mL
refractive index   n20/D 1.444(lit.)
bp   60.9 °C(lit.)
mp   −64 °C(lit.)
density   1.500 g/mL at 25 °C(lit.)
mass shift   M+1

Price and Availability

Safety & Documentation
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

accessory

Product #

Description

Add to Cart

Z693847 Scienceware® Break-Safe ampule opener

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?