EMAIL THIS PAGE TO A FRIEND
442887 Aldrich

Pinacolone oxime

98%

Synonym: 3,3-Dimethyl-2-butanone oxime

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Nitrogen Compounds, Organic Building Blocks, Oximes More...
InChI Key   UNSDDJQNHCSVSW-FNORWQNLSA-N
assay   98%
mp   76-78 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?