• USA Home
  • 443670 - Butane, fuel for Micro Torch

EMAIL THIS PAGE TO A FRIEND
443670 Aldrich

Butane, fuel for Micro Torch

contains no CFC gases

Synonym: butane fuel, butane gas, gas butane

Purchase

Properties

Related Categories Burners, Heating, Labware More...
vapor density   2.11 (vs air)
vapor pressure   51.6 psi ( 37.7 °C)
InChI Key   IJDNQMDRQITEOD-UHFFFAOYSA-N
autoignition temp.   761 °F
expl. lim.   8.4 %
packaging   pkg of 12 × 26 g
availability   not available in EU
bp   −0.5 °C(lit.)
mp   −138 °C(lit.)

Description

General description

• Triple-refined formula reduces clogging of filters
• Special vapor pressure promotes consistent ignition
• Contains no CFC’s, canister includes single-piece universal filling tip
• Recommended refill for all Ultratorch® & Master Butane-powered products
• 26g of butane fuel per can

Legal Information

Ultratorch is a registered trademark of Master Appliance Corp.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2037F 2.1
RTECS 
EJ4200000
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?