• USA Home
  • 444766 - Dimethyl sulfoxide-d6

EMAIL THIS PAGE TO A FRIEND
444766 Aldrich

Dimethyl sulfoxide-d6

"100%", 99.96 atom % D

Synonym: (Methyl sulfoxide)-d6, DMSO-d6, Hexadeuterodimethyl sulfoxide

Purchase

Properties

Related Categories Dimethyl sulfoxide-d6, NMR Solvents, Solvent by Application, Solvents, Spectroscopy Solvents (IR, NMR, UV/Vis) More...
vapor pressure   0.42 mmHg ( 20 °C)
InChI Key   IAZDPXIOMUYVGZ-WFGJKAKNSA-N
isotopic purity   99.96 atom % D
autoignition temp.   573 °F
expl. lim.   42 %
packaging   pk of 10 × 0.75 mL
refractive index   n20/D 1.476(lit.)
bp   189 °C(lit.)
density   1.190 g/mL at 25 °C(lit.)
mass shift   M+6

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NA 1993 / PGIII
WGK Germany 
2
Flash Point(F) 
188.6 °F
Flash Point(C) 
87 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

accessory

Product #

Description

Add to Cart

Z693847 Scienceware® Break-Safe ampule opener

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?