EMAIL THIS PAGE TO A FRIEND
444782 Aldrich

1-Bromododecane

95%

Synonym: Dodecyl bromide, Lauryl bromide

Purchase

Properties

vapor density   8.6 (vs air)
InChI Key   PBLNBZIONSLZBU-UHFFFAOYSA-N
assay   95%
refractive index   n20/D 1.458(lit.)
bp   134-135 °C/6 mmHg(lit.)
mp   −11-−9 °C(lit.)
density   1.038 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
WGK Germany 
3
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?