EMAIL THIS PAGE TO A FRIEND
445002 Aldrich

Fmoc-D-Trp-OH

98%

Synonym: Nα-(9-Fluorenylmethoxycarbonyl)-D-tryptophan, Fmoc-D-tryptophan

  • CAS Number 86123-11-7

  • Empirical Formula (Hill Notation) C26H22N2O4

  • Molecular Weight 426.46

  •  Beilstein Registry Number 5462683

  •  MDL number MFCD00062954

Purchase

Properties

InChI Key   MGHMWKZOLAAOTD-XMMPIXPASA-N
assay   98%
optical activity   [α]20/D +28°, c = 1 in DMF
mp   182-185 °C(lit.)

Description

Application

Cyclic peptides used as tachykinin antagonists have been prepared from Fmoc-protected D- and L- tryptophans.

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Related Content

Indoles, Purines, and Their Isosteres

Substituted indoles and purines have frequently been referred to as “privileged structures” since they are capable of binding to multiple receptors with high affinity, and thus have applications acro...
Keywords: Building blocks, Medicinal chemistry

Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?