• USA Home
  • 445010 - N-(9-Fmoc)-L-glutamic acid γ-tert-butyl ester monohydrate

EMAIL THIS PAGE TO A FRIEND
445010 Aldrich

N-(9-Fmoc)-L-glutamic acid γ-tert-butyl ester monohydrate

98%

Synonym: N-(9-Fluorenylmethoxycarbonyl)-L-glutamic acid γ-tert-butyl ester monohydrate

  • CAS Number 204251-24-1

  • Empirical Formula (Hill Notation) C24H27NO6 · H2O

  • Molecular Weight 443.49

  •  MDL number MFCD00150485

  •  PubChem Substance ID 24868156

Purchase

Properties

Related Categories Amino Acids & Derivatives, Chemical Biology, Chemical Synthesis, Glutamic Acid, Peptide Chemistry More...
InChI Key   NMBGBVUJSPZRDD-BDQAORGHSA-N
assay   98%
optical activity   [α]20/D −17.5°, c = 1 in DMF
mp   86-89 °C(lit.)
storage temp.   2-8°C

Description

Packaging

1 g in glass bottle

Application

Protected building block for the introduction of glutamic acid into peptides.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?