• USA Home
  • 445053 - Ethyl cis-5-dodecenoate

EMAIL THIS PAGE TO A FRIEND
445053 Aldrich

Ethyl cis-5-dodecenoate

98%

  • CAS Number 87028-10-2

  • Empirical Formula (Hill Notation) C14H26O2

  • Molecular Weight 226.36

  •  MDL number MFCD00239342

Purchase

Properties

Related Categories Building Blocks, C12 to C63, Carbonyl Compounds, Chemical Synthesis, Esters,
InChI Key   LYMJBMIYQMVCCY-KTKRTIGZSA-N
assay   98%

Price and Availability

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
>235.4 °F
Flash Point(C) 
>113 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?