• USA Home
  • 445975 - Dow Corning® silicone diffusion pump fluids

EMAIL THIS PAGE TO A FRIEND
445975 Aldrich

Dow Corning® silicone diffusion pump fluids

704

Purchase

Properties

Related Categories Labware, Pumps, Vacuum Pump Accessories, Vacuum Pumps
InChI Key   YFCVAZGXPLMNDG-UHFFFAOYSA-N
refractive index   n20/D 1.559
bp   215 °C/0.5 mmHg
density   1.07 g/mL at 25 °C

Description

Application

Single component fluid for high vacuums of 10-6 to 10-8 torr (untrapped) and 10-10 to 10-11 torr (trapped). Offers quick pumpdown.

General description

For high vacuum and for fast pumping of large volumes of gas or vapor in production operations. Chemically and thermally inert fluids. General purpose for pumping large volumes of gas, vacuum range 10-5 to 10-7 torr. Also suitable for use in Aldrich swivel McLeod vacuum gauges.

Legal Information

Dow Corning is a registered trademark of The Dow Chemical Company or an affiliated company of Dow

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
WGK Germany 
3
Flash Point(F) 
213.8 °F
Flash Point(C) 
101 °C
Protocols & Articles

Articles

Aldrich 3-Stage Oil Diffusion Pump

Aldrich 3-stage oil diffusion pumps provide fast 375 L/s pumping speed that outperforms traditional glass diffusion pumps. They feature all glass construction with a removable jet assembly for easy c...
Chemfiles Volume 4 Article 3
Keywords: Catalog, Diffusion, Solvents, Sublimation, Type

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?